Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromosuccinic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: 2,3-Dibromobutanedioic acid

M-Clarity™ quality level = MQ200

8.41348.0050 8.41348.0050
EM8.41348.0050CS 11651 USD InStock
CA8.41348.0050 EM8.41348.0050
2,3-Dibromosuccinic acid for synthesis, Sigma-Aldrich®
2,3-Dibromosuccinic acid


Formula: HOOC(BrCH)₂COOH
MW: 275.88 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 278…281 °C
MDL Number: MFCD00004209
CAS Number: 526-78-3
EINECS: 208-396-9
UN: 3261
ADR: 8,III
Merck Index: 12,03078
Order Now
Specification Test Results
Melting Point 278 - 281 °C
Molar Mass 275.88 g/mol

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty