Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thiosalicylic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Thiobenzoic acid2-Mercaptobenzoic acid

M-Clarity™ quality level = MQ200

8.41613.0100 8.41613.0100
EM8.41613.0100CS 10292 USD InStock
CA8.41613.0100 EM8.41613.0100
Thiosalicylic acid for synthesis, Sigma-Aldrich®
Thiosalicylic acid


Formula: C₇H₆O₂S
MW: 154.19 g/mol
Boiling Pt: 230 °C (0.02 mmHg)
Melting Pt: 165…166 °C
Density: 1.489 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004836
CAS Number: 147-93-3
EINECS: 205-704-3
ADR: 9,
Merck Index: 12,09498
Order Now
Specification Test Results
Solubility (20 °C) practically insoluble
Melting Point 165 - 166 °C (sublimed)
Molar Mass 154.19 g/mol
Density 1.49 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty