Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxy-4-vinylphenol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning
Synonyms: 2-Hydroxy-5-vinylanisole

M-Clarity™ quality level = MQ200

8.43803.0025 8.43803.0025
EM8.43803.0025CS 37638 USD InStock
EM8.43803.0025 CA8.43803.0025
2-Methoxy-4-vinylphenol for synthesis, Sigma-Aldrich®
2-Methoxy-4-vinylphenol


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
MDL Number: MFCD00015437
CAS Number: 7786-61-0
EINECS: 232-101-2
Order Now
Specification Test Results
Melting Point 9 - 10 °C
Molar Mass 150.18 g/mol
Boiling Point 224 °C (1013 hPa)
Density 1.11 g/cm3 (14 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty