Would you like to log in again?
Rubbermaid® Commercial Triple Trolley
Supplier: Rubbermaid
Rubbermaid®
RCP4401BLA
19757-224EA 1131.86 USD InStock
19757-224
Rubbermaid® Commercial Triple Trolley
Carts
The Rubbermaid Commercial Triple Trolley efficiently transports large bulky loads and then folds flat for easy storage.
  • Handle easily converts trolley to dolly
  • Plastic material is easy on walls and floors
  • With one shelf
  • Weight capacity of 500 lbs.
The trolley has a sturdy structural foam deck that won't rust, dent, chip, or peel. The 3-position handle allows the user to push, pull, or fold flat for use as a dolly. Tough plastic construction is easy on walls and doors. The 5-inch casters are non-marking.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty