Would you like to log in again?
Rubbermaid® Commercial Microfiber Dust Pads
Supplier: Rubbermaid
HYGEN™
RCP Q412 WHI RCPQ418GNCT
19758-830CS 493.61 USD InStock
19758-830 89128-094
Rubbermaid® Commercial Microfiber Dust Pads
Mops
The Rubbermaid Commercial Products HYGEN Microfiber Dust Mop is a premium fringed floor mop designed to remove dust and help prevent the spread of infection.
  • Durable for up to 500 launderings (up to 200 with bleach)
  • Bleach-safe in hot or cold water
  • Double finished edges retain mop shape with use over time
  • Washable
These floor mops are made from 100% microfiber to trap and lock away dust, dirt and debris. These floor mop pads are safe for laundering, even with bleach in hot or cold water.
Its fringe provides greater surface area for better debris pickup and faster cleaning. Fringed Microfiber Dust pad grabs and holds larger dust particles and debris, providing 50% more surface area than non-fringed pads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty