Would you like to log in again?
Stainless Steel Scoop, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
885540
66174-540CS 216.57 USD InStock
66174-540
Stainless Steel Scoop, Wheaton, DWK Life Sciences
Scoops
This scoop is made from 302 stainless steel and is ideal for sampling powders, slurries, soils and more
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty