Would you like to log in again?
PHC3081-8 Combination pH Electrode with Temperature Sensor, Radiometer Analytical, Hach
Supplier: Hach
E16M305 E16M305
89501-024EA 932.41 USD InStock
CA89501-024 89501-024
PHC3081-8 Combination pH Electrode with Temperature Sensor, Radiometer Analytical, Hach
Electrodes pH Electrodes
The Radiometer PHC3081-8 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat
Glass sensors made for long-life and durable use.
AgCl), integrated temperature sensor, and porous pin junction for general purpose applications. The PHC3081-8 has BNC + cinch (RCA) connectors. Please reference the PHC3081-6 for a version of this probe compatible with Hach sensION+ benchtop pH meters.
Ordering information: Includes Radiometer Analytical PHC3081-8 combination pH electrode with Certificate of Conformity and small bottle of internal electrolyte solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty