Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Bel-Art Teflon® Policeman, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H37738-0000
13197-416CS 381 USD InStock
13197-416
SP Bel-Art Teflon® Policeman, Bel-Art Products, a part of SP
Spatulas
Made of a 7.9 mm (⁵/₁₆) Ø Teflon® PTFE rod with flattened ends, this policeman is useful for scraping and transferring materials.
  • Useful for scraping and transferring materials
  • Durable PTFE construction
  • Flattened ends
  • Autoclavable
Steam autoclavable at 121 °C (250 °F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty