Would you like to log in again?
Mucasol® Universal Detergent, BrandTech® Scientific
Supplier: BrandTech
Mucasol®
44003B
89089-400EA 360.06 USD InStock
89089-400
Mucasol® Universal Detergent, BrandTech® Scientific
Cleaning Agents Detergents and Surface Cleansers
An excellent alternative to chromosulfuric acid, Mucasol® laboratory detergent is an alkaline, high-performance liquid concentrate for wiping down lab surfaces and cleaning labware in immersion and ultrasonic baths.
  • Laboratory detergent
  • Highly concentrated
  • Excellent for glassware
  • Alternative to chromo-sulfuric acid
  • Suitable for radioactive decontamination
Excellent for a wide range of lab surfaces. Use Mucasol lab detergent for cleaning glass, porcelain, plastic, rubber, and metal labware. Prolonged exposure to zinc and non-anodized aluminum should be avoided.
Suitable for radioactive decontamination. Mucasol is bacteriostatic and suitable for radioactive decontamination.
One bottle provides 1,000L of cleaning solution. Mucasol is concentrated and formulated for use at concentrations between 0.2 and 3%. Just a single two-liter bottle can produce up to 1,000L of cleaning solution.
Mucasol lab detergent features a formula that is gentle to the skin, non-toxic, non-aggressive, and biodegradeable.
Ordering information: Please note that Mucasol lab detergent contains phosphates; check local regulations prior to use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty