Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACTA2 Monoclonal Antibody [clone: 5C5]

Supplier: Diagnostic BioSystems

PDM099 MOB128-05 MOB128
95024-216EA 401.19 USD InStock
95024-216 99990-328 99990-978
Anti-ACTA2 Monoclonal Antibody [clone: 5C5]
Antibodies
Monoclonal anti-α-sarcomeric actin has been used as a marker for rhabdomyosarcoma. This antibody is specific to α-skeletal and α-cardiac muscle actins. It does not react with smooth muscle tissue.

Type:
Antigen: ACTA2
Clonality: Monoclonal
Clone: 5C5
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty