Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Anti-C4C Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Diagnostic BioSystems

F005
99990-038EA 521.67 USD InStock
99990-038
Anti-C4C Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This antibody reacts with human C4, C4b, and C4c but does not react with the C4d epitope. Traces of contaminating antibodies have been removed by solid phase absorption with human plasma proteins. This antibody is well suited for tissue section staining but may also be used for other applications.

Type: 0
Antigen: C4C
Clonality: 0
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: 0
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty