Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD74 Lymphoid Marker Mouse Monoclonal Antibody [clone: LN2]

Supplier: Diagnostic BioSystems

MOB168 PDM033 MOB168-05
99990-372EA 560.96 USD InStock
99990-372 95024-378 99990-876
Anti-CD74 Lymphoid Marker Mouse Monoclonal Antibody [clone: LN2]
Antibodies
This antibody is specific to CD74. This antibody can be used for the identification of B cell lymphomas and leukemias. It also stains activated neoplastic cells in T cell lymphomas and Sternberg-Reed cells.

Type:
Antigen: CD74 Lymphoid Marker
Clonality: Monoclonal
Clone: LN2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty