Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B3GAT1 Mouse Monoclonal Antibody [clone: NK-1]

Supplier: Diagnostic BioSystems

PDM130 MOB163 MOB163-05
99990-364EA 619.79 USD InStock
99990-364 95024-368 99991-032
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: NK-1]
Antibodies
This antibody is specific to a 110kDa human CD57 myeloid associated glycoprotein. It stains mononuclear cells with natural killer activity, neuroendocrine cells and their tumors.

Type:
Antigen: B3GAT1
Clonality: Monoclonal
Clone: NK-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty