Would you like to log in again?
DeBakey Atraumatic Bulldog Clamp, Roboz Surgical
Supplier: Roboz
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
RS-7511 RS-7500
100188-880EA 437.54 USD InStock
100188-880 100188-886
DeBakey Atraumatic Bulldog Clamp, Roboz Surgical
Clamps General Purpose Clamps
The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty