Would you like to log in again?
Human Peripheral Blood Natural Killer (NK) Cells, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB56C-1 PB56NC-2 PB56C-2 PB56NC-1
101190-026EA 1730.77 USD InStock
101190-026 101190-024 101190-084 101190-086
Human Peripheral Blood Natural Killer (NK) Cells, Charles River Laboratories Cell Solutions
Blood
Human peripheral blood CD56 NK Cells are isolated from peripheral blood mononuclear cells (PBMCs) using positive or negative immunomagnetic cell separation procedures (upon your request)
PBMCs are collected in HemaCare's FDA-registered collection centers following cGMP and cGTP collection guidelines from healthy human volunteer donors under IRB approved informed consent by leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol. This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques. Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
CD56 NK cells activity has recently been a focus of cancer research due to the cell type’s natural ability to seek out and destroy abnormal cells without any kind of MHC signal. Studies have shown that NK cells referred to as lyphikine-activated killers (LAK), can be induced by higher than normal IL-2 levels to increase their cytotoxic activity towards cell lines, like many cancer cells, initially resistant to un-stimulated NK cells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty