Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Anti-CK7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]

Supplier: Diagnostic BioSystems

PDM097 MOB057 MOB057-05 MOB057-05 PDM097 MOB057
95024-456EA 351.51 USD InStock
95024-456 99990-974 99990-226 CA95024-456 CA99990-974 CA99990-226
Anti-CK7 Mouse Monoclonal Antibody [clone: OV-TL 12/30]
Antibodies
This antibody recognizes keratin polypeptide of 54 kDa. It stains a number of epithelial cells including ductal and glandular epithelia. Stratified squamous epithelia are positive but transitional epithelium of the urinary tract does not show any staining. This antibody labels adenocarcinomas and transitional carcinomas but does not react with squamous cell carcinomas.

Type:
Antigen: CK7
Clonality: Monoclonal
Clone: OV-TL 12/30
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty