Would you like to log in again?
Wooden Tweezers Holder Case, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72898-01
102098-074EA 100.64 USD InStock
102098-074
Wooden Tweezers Holder Case, Electron Microscopy Sciences
Boxes Storage Boxes
Made from solid wood, with brass double hinges and an elegant latch.
  • Holds 5 tweezers with a maximum length of 140 mm
Delivery information: Supplied with a foam padded top and bottom to protect stored tweezers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty