Would you like to log in again?
Dumont Tweezers Style N7AC, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0203-N7AC-PO 0202-N7AC-PO
102091-594EA 103.18 USD InStock
102091-594 102091-574
Dumont Tweezers Style N7AC, Electron Microscopy Sciences
Forceps
Anti-capillary, type 7, super thin tips. Length: 115 mm.
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. lnox 02 will resist temperatures of around 400°C (DIN 50 914), but cannot be sterilized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty