Would you like to log in again?
1 Gal Paraffin Dispenser, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62839-10 62839-20
101412-656EA 7716.92 USD InStock
101412-656 101412-654
1 Gal Paraffin Dispenser, Electron Microscopy Sciences
Paraffin Wax Trimmers and Dispensers
This dispenser provides immediate 'Wax on Tap' for use in pathology laboratories. Durable construction of an anodized black interior stove, enameled black and white aluminum exterior and a black metal lid.
  • Thermally insulated tap heater with independent temperature contol eliminates wax solidification.
  • Built in controller provides temperature control from ambient to 75 °C
  • A non-drip swivel lever with replaceable filter makes the dispensing of wax easy
  • Safety over-temperature at 105 °C; non-resettable thermal fuse
  • Easy to clean
Capacity of 1 gal. (4.5 liters)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty