Would you like to log in again?
Silver Conductive 18DB70X, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12684-15
100496-456EA 102.55 USD InStock
100496-456
Silver Conductive 18DB70X, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Silver Conductive Coating 18DB70X is a direct substitute for Silver Conductive Fluid 416, which is no longer available.
  • Low VOC
  • Low ohms
  • Excellent long term shielding and grounding properties
This material was specially formulated for use in geographic areas that demand the use of low VOC (Volatile Organic Compounds) products. It also provides low ohms at very thin dry film thickness on almost any surface, and exhibits excellent long term shielding and grounding properties.
Certifications: Certificates of analysis are available from Electron Microscopy Sciences. Please contact us or visit our website to search by lot or product number.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty