Would you like to log in again?
Alcotabs® Effervescent Tablets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Alcotabs®
60530 60529
100504-778CS 484.96 USD InStock
100504-778 100504-836
Alcotabs® Effervescent Tablets, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Alconox detergents in an effervescent tablet form for use in syphon type washer rinsers for the cleaning of pipettes and test tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty