Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Araldite 502, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Araldite®
13900
100503-422KT 81.8 USD InStock
100503-422
Araldite 502, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Luft's Formula.
  • Araldite 502 Kit
  • Polymerization takes place overnight
  • Blocks can be sectioned the next day
Polymerization takes place overnight so blocks can be sectioned the next day
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, that in order to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5-2%; BDMA: 2.5-3%).
J. Biochem. Biophy. Cytol. 9, 409 (1961) Kit consists of: 450 mL Araldite 502, 450 mL DDSA; Specially Distilled, and *50 mL DMP-30.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty