Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Araldite, Embed 812, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Araldite®
13940
100503-426KT 139.84 USD InStock
100503-426
Araldite, Embed 812, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Mollenhauer Epon-Araldite Formula. Sectioning with this mixture of resins has been proven to be easier than with each individual resin and the thermal stability of araldite and the image contrast of EM-bed-812 have made this a very successful blend.

  • For hard blocks
  • Blocks are easily sectioned
  • Developed for embedding plant tissue
  • Also successful with animal tissue

For hard blocks and high image contrast

*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5 to 2%; BDMA: 2.5 to 3%).

Ordering information: Kit consists of: 450 mL EMbed-812, 450 mL Araldite 502, 450 mL DDSA; Specially Distilled, *50 mL DMP.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty