Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Beryllium, support films

Supplier: Electron Microscopy Sciences

76030
100492-290EA 3557.08 USD InStock
100492-290
Beryllium, support films
Beryllium
A deposition of 250 Angstroms thick Beryllium onto the 0.005" thick, 25 x 25 mm squares of a Cu substrate. The Be can be removed by dissolving the substrate in nitric acid (50:50). The Be film will then be removed from the acid, washed in distilled water and mounted on TEM grids. A Be support film will reduce background interference to a minimum and it is particularly useful where analyses for C or Si are required, so that these alternative supports cannot be used. Another advantage of the Be support is its very fine grain size which produces a very sharp ring pattern for in-situ calibration.


Formula: Be
MW: 9.01 g/mol
MDL Number: MFCD00134032
CAS Number: 7440-41-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty