Would you like to log in again?
Catalase Crystals
Supplier: Electron Microscopy Sciences
80014
100501-376EA 174.92 USD InStock
100501-376
Catalase Crystals
Enzymes
Catalase crystals are mounted on a grid (3.05 mm) and negatively stained.
  • Ideal for high magnification calibration
  • They display very clear lattice plane spacings of approx. 8.75 and 6.85 nm
J. Ultrastructure Res.24, 454., 1968
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty