Would you like to log in again?
CCC Carbon Adhesive, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12664 12665
100503-782EA 25.58 USD InStock
100503-782 100503-780
CCC Carbon Adhesive, Electron Microscopy Sciences
Stains Microscopy and staining reagents
An electrically Carbon Conductive Cement for specimen mounting in all SEM work.
  • For specimen mounting
  • Use in SEM work
  • Thinner is available
After drying of the cement, immediate investigation of conductive specimens is possible. Non-conductive specimens need only to be coated with carbon or metal. Thinner is available if the cement viscosity is too thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty