Would you like to log in again?
Chessy Test Specimen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
79503-01
100494-944EA 4393.34 USD InStock
100494-944
Chessy Test Specimen, Electron Microscopy Sciences
Specimens
A precise SEM test specimen for most all calibration applications. The edges of the empty corners in 0.1 and 1 mm checkerboards are additionally marked
  • Resolution test at high magnification on the edges of the gold squares
  • The pattern was directly written by e-beam lithography using the new ZBA 31/32 from JENOPTIK
There are more than 1.6 million gold squares of 1µ size on silicon forming a 4-fold checkerboard pattern in an area of 5 mm square. The smallest metric checkerboard has a size of 10×10 µm. Such checkerboards form larger metric checkerboards of 100×100 µm. These again form checkerboards of 1 mm square. Finally, such 1 mm squares are arranged in the same manner covering a field of 5 mm square. The surrounding frame is 10 µm wide and has an outer side length of 5.04 mm.
Imaging for calibration of SEM magnification in all ranges between 20× and 50000×. Check for equal scaling in X and Y and also for orthogonality and distortion.
Motorized stages: (especially for use with ESCOSY) Measurement of reproducibility using stored position and absolute positioning accuracy and SEM distortion at any magnification via mark recognition on different places. Calibration of readings in X and Y. Calibration of stage orthogonality. Experimental Electron Lithography (use with attachment ELPHY or PROXY-WRITER). Generation of metric writing fields between 10 µ and 5 mm square via mark recognition and alignment. Check of defocusing in outer areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty