Would you like to log in again?
Conductive Silver Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12644-01 12644-02
100496-450EA 93.21 USD InStock
100496-450 100496-448
Conductive Silver Pen, Electron Microscopy Sciences
Stains Microscopy and staining reagents
This pen is designed for making instant conductive silver traces.
  • Unique valve tip allows for very smooth flow
  • Spring loaded to prevent clogging
  • Approximately 100 feet of traces per pen
Pen is ideal for applying samples to SEM stubs. A unique valve tip allows for very smooth flow with normal writing pressure and it is spring loaded to prevent clogging. For conductivity traces, solderable terminations are possible using a 250°F cure for 15-20 minutes. Tin, lead, or silver solder can be used (Do not exceed 350°F for more than 5 seconds). Each pen is filled with approximately 100 feet of traces. Silver content: 39-45% with less than 10 micron diameter. The thinner that is used for this pen is Butyl Acetate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty