Would you like to log in again?
Plunge Freezer - EMS-002 Rapid Immersion Freezer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
37000
100496-104EA 22159.39 USD InStock
100496-104
Plunge Freezer - EMS-002 Rapid Immersion Freezer, Electron Microscopy Sciences
Freezers Laboratory / General Purpose Freezers
Ultrarapid freezing, when used in conjunction with appropriate accessory processing methods has long been recognized to provide reservation of ultrastructure, localization of soluble inorganic and organic species, and preservation of antigenicity that is superior to that obtainable through conventional chemical fixation techniques and the use of cryoprotectants. Furthermore, it is the only procedure currently available for capturing events that occur on timescales less than a few seconds. Successful ultrarapid freezing requires that heat be withdrawn so rapidly from an unfixed un-cryoprotected sample that no ice crystal damage is visible at the electron microscopic level of analysis. The EMS-002 Cryo Workstation allows for a reliable virtually routine freezing of biological samples and materials.
  • Removable Drop Anvil Assemble
  • Specimen Holder
  • Stainless Steel Liquid Nitrogen Dewar
  • Cryogen Temperature Controller
  • Drop Release
  • Removable Cryogen Holder for safe disposal of Cryogen
  • Frozen Specimen Work Station
  • Economical, universal application rapid freezing unit
  • Plunge freezing with temperature controlled cryogen
  • Frozen samples handled under cold dry nitrogen atmosphere
  • Rapid set up and easy use
  • Reproducible results
  • High and low set points for cryogen control
The EMS-002 achieves ultra-rapid cooling by quenching the sample in liquid propane or other suitable cryogen, cooled to near liquid nitrogen temperature
The device requires only about 10 minutes to cool to operating temperature after the first filling of propane, and less than five minutes after subsequent fillings. The operator can increase or decrease the rate of cooling by adjusting the level of liquid nitrogen in the Dewar. Heaters built into the chamber wall, powered by the temperature controller, evenly warms the propane above its melting point. The controllers’ temperature is set for the kind of cryogen being used, for instance, 185°C for commercial grade propane. Chemically pure propane (map. 190°C) can also be used, but it is significantly more expensive than commercial grade propane and confers no particular advantage. Ethane and a non-flammable cryogen, SUVA-124 produced by Dupont can also be used as a cryogen in this instrument. When using flammable cryogens like Propane & Ethane we recommend using the EMS-002 in a fume hood.
The EMS-002 accomplishes this by using a stainless steel vacuum insulated Dewar and copper condensation chamber. Liquid nitrogen is poured into the Dewar to a level determined by the user. The initial cooling of the cryogen condensation chamber takes about 15 to 20 minutes. Gaseous propane is then admitted to the condensation chamber slowly enough to allow it to condense and fill the chamber with liquid propane. The propane is led to cool to near liquid nitrogen temperature supervised by the digital temperature controller while the sample is being prepared for freezing.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty