Would you like to log in again?
Desiccant in a Bag, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71209-20 71209-25 71209-16
100502-824EA 292.9 USD InStock
100502-824 100493-142 100493-144
Desiccant in a Bag, Electron Microscopy Sciences
Desiccators Desiccants
The desiccant regenerates repeatedly without degradation in a 150 °C (300 °F) vented oven for 3 hours or more (depending on size).
  • Indicating gel beads change color from blue when dry to pink when moist
  • Dry until the blue color is restored
Packaging: Supplied in bags with viewing area which allows user to know when to recharge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty