Would you like to log in again?
Diethylene Glycol Distearate (DEGDS/DGD), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
14010
100503-430KT 307.83 USD InStock
100503-430
Diethylene Glycol Distearate (DEGDS/DGD), Electron Microscopy Sciences
Stains Microscopy and staining reagents
DGD is a removable embedding medium (for thin and thick sections) that provides embedment free sections
  • Thin or thick embedment free sections can be obtained
  • Sections are easy to cut and ribbons are easily obtainable
  • Enhances the integrity of the specimen as it floats on the water
  • Sections produce interference colors according to its thickness
  • Ethanol and n-butyl alcohol are used as dehydrating agents
Kit consists of: 450 mL DGD, 450 mL Ethanol, 450 mL n-Butyl alcohol, and DGD Embedding Kit.
Good for immunolabeling and high resolution light microscopy.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty