Would you like to log in again?
Disposable Charcoal Cartridge, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64454-05
100490-582PK 109.13 USD InStock
100490-582
Disposable Charcoal Cartridge, Electron Microscopy Sciences
Culture Dishes Cell Culture Dishes
Absorb Incubator Contaminants. This disposable, activated charcoal cartridge efficiently absorbs low energy 35S radioactive components during metabolic labeling. The containment problem exists in incubators and is caused by 35S-labeled amino acids, such as methionine and cysteine which are used to label proteins. The charcoal is supplied in a 100mm diameter plastic petri dish with a highly porous membrane sealed to the top edge.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty