Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

E2M1.5 Vacuum Pump, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
91004 91004-E
100492-476EA 6081.36 USD InStock
100492-476 100495-458
E2M1.5 Vacuum Pump, Electron Microscopy Sciences
Pumps Vacuum Pumps
This miniature two stage pump features an alternative inlet connection position at the side of the pump so that overall installation height can be reduced to a minimum when required.

  • 1.3 cfm (190L/min) displacement
  • Ultimate vacuum (without gas ballast): 1.1 x 10-3 torr
  • Max. inlet pressure for water vapor: 11.3 torr
  • Max. water vapor pumping rate: 0.035 lb/hr
  • Oil capacity Max/Min: 0.28L/0.2L

Ultra Grade 19 oil is recommended for use with the E2M1. Weighs 50 lbs (22kg).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty