Would you like to log in again?
Edwards Ultragrade 19 Oil, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
60742
100504-368EA 178.44 USD InStock
100504-368
Edwards Ultragrade 19 Oil, Electron Microscopy Sciences
Lubricants
Edwards Ultragrade 19 Oil is specially recommended for use in clean pumping applications
The oil offers reduced maintenance cost, longer fluid life, and a cooler running pump.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty