Would you like to log in again?
EMbed 812 Embedding Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14120
100503-438KT 164.76 USD InStock
100503-438
EMbed 812 Embedding Kit, Electron Microscopy Sciences
Stains Microscopy and staining reagents
The same advantages offered by EPON 812, are offered by EMbed 812: rapid penetration, greater contrast, easy sectioning, stability under the electron beam, satisfactory staining of most thick sections for light microscopy and thin sections for electron microscopy.
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, that in order to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5 to 2%; BDMA: 2.5 to 3%).
Delivery information: Kit consists of 450 mL EMbed-812, 450 mL DDSA; Specially Distilled, 450 mL NMA, *50 mL DMP-30.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty