Would you like to log in again?
MicroPoint™ Micro Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62085-3S 62086-3S 62085-2S 62086-2S 62086-1S 62085-1S
100497-618EA 128.53 USD InStock
100497-618 100497-620 100497-622 100497-624 100497-614 100497-616
MicroPoint™ Micro Forceps, Electron Microscopy Sciences
Forceps
Micro Forceps MF-1, Straight, Smooth or Serrated Jaws features extra delicate tips for general precision work
Micro Forceps/Tweezers are designed specifically for applications in the field of microscopic science.
Micro Forceps MF-1, Straight, Smooth or Serrated Jaws features extra delicate tips for general precision work. 100 mm (4.0”) in length, tips 0.5 mm wide.
Micro Forceps MF-2, Slightly Curved. Smooth or Serrated Jaws especially designed for precise handling in areas of limited access. Very delicate. 100 mm (4.0”) in length; 0.5 mm width in tips.
Micro Forceps MF-3, Fully Curved. Smooth or Serrated Jaws is designed for optimum visibility in precision microscopic work. Very delicate. 100 mm (4.0”) in length; 0.5 mm width in tips
100 mm (4.0”) in length, tips 0.5 mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty