Would you like to log in again?
Roll Film Washer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74386
100492-190EA 112.44 USD InStock
100492-190
Roll Film Washer, Electron Microscopy Sciences
Film Processing Supplies
For complete washing of your roll film. The vacuum action will gently wash your film to archival standards. This conventional overflow washer lets water in from the bottom and the contaminated water out from the top. The vacuum action washer fills the water from the bottom, but quickly and automatically empties from the bottom by a vacuum syphon action, which discharges all contaminated water 2-3 times per minute. The vacuum action washer also protects your film from abrasions caused by high turbulance and unwinding of the reel, which happenes in conventional washers. Holds 5: 35 mm reels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty