Would you like to log in again?
Slide Tray for 100 Slides , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71511-10 71510-01 71510-10 71511-01
100491-000PK 782.6 USD InStock
100491-000 100493-386 100493-388 100501-310
Slide Tray for 100 Slides , Electron Microscopy Sciences
Slide Holders
Made from tough, resilient ASB plastic which offers long life.
Holds 100-standard 1 x 3" slides positioned vertically for easy insertion in pre-numbered grooves allowing for simple identification and retrieval. Dimensions L x W x H: 14.25 x 0.25 x 3.75". A cover which is optional is made from clear polystyrene offering full visibility and it protects the slides from dirt and dust when they are not in the drawer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty