Would you like to log in again?
Cellattice™ Micro-Ruled Plastic Cell Culture Surfaces, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63571-25
102094-792PK 182.94 USD InStock
102094-792
Cellattice™ Micro-Ruled Plastic Cell Culture Surfaces, Electron Microscopy Sciences
Specimen Mounting Supplies
Measure the Same Cell: Cell cycle, axon growth and differentiation
Direct Cell Count: Proliferation and cytotoxicity without removing attached cells
  • Measure the Same Cell: Cell cycle, axon growth and differentiation
  • Direct Cell Count: Proliferation and cytotoxicity without removing attached cells
The built-in markers on the Cellattice allow direct measurement of cell growth and movement without image acquisition
Cellatice™ can be used to monitor morphology, cell movement and differentiation at the individual cell level. Cell proliferation is measured directly with multiple reading.
Has a grid area of 10 x 10 mm
Same cell or cell clusters can be measured throughout the time course of the experiment, even after each incubation period.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty