Would you like to log in again?
Leit-C-Plast Adhesive Material, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12667
100503-784EA 110.35 USD InStock
100503-784
Leit-C-Plast Adhesive Material, Electron Microscopy Sciences
Stains Microscopy and staining reagents
A special adhesive with very high electrical conductivity and permanent plasticity.
  • Very high electrical conductivity
  • Permanent plasticity
Use for the preparation of big specimens in SEM work.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty