Would you like to log in again?
LR Gold Resin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14371 14370
100504-910KT 770.88 USD InStock
100504-910 100503-860
LR Gold Resin, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Kit consists of 500 mL LR Gold Resin, 100g Polyvinylpyrolidone; 50g Benzoyl Peroxide, and 50g Benzil.
The London Resin Company's acrylic "LR Gold" offers the histochemist and immunocytochemist the advantage of working with unfixed tissues at low temperatures, thus allowing many fixation-sensitive procedures to be carried out in 1-4 microns resin sections at -25°C using visible light. It is non-toxic and has a low viscosity.
Kit consists of 500 mL LR Gold Resin, 100g Polyvinylpyrolidone; 50g Benzoyl Peroxide, and 50g Benzil.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty