Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Negative Carriers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74868-45 74867-10 74868-10 74868-20 74867-40 74867-35 74868-40
100494-098EA 37.43 USD InStock
100494-098 100494-088 100494-096 100494-086 100494-072 100494-084 100494-082
Negative Carriers, Electron Microscopy Sciences
Film Processing Supplies
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts
Glassless, sandwich type negative carriers are rofessional quality, with black, anodized aluminum construction. Hinged for quick handling.
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts. Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty