Would you like to log in again?
Aurion Normal Serum, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
25568 25578 25572 25570
100487-952EA 80.27 USD InStock
100487-952 100487-944 100487-948 100487-946
Aurion Normal Serum, Electron Microscopy Science
Sera (Serum)
Normal sera are used to counteract the non-specific interaction between the sample and immunoglobulins. They can be added to the blocking solution and the incubation media.
As a rule the normal serum species should be the same as the secondary antibody species (use normal goat serum with goat-anti-rabbit conjugates).
Note: normal sera should not be used in combination with Protein A and Protein G gold reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty