Would you like to log in again?
Plastic Forceps, with Guide Pin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72970
100488-492PK 27.6 USD InStock
100488-492
Plastic Forceps, with Guide Pin, Electron Microscopy Sciences
Forceps
Forceps are 12.7 cm (5") long, molded, strong, durable, autoclavable up to 275°F (135°C), serrated tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty