Would you like to log in again?
Plastic Forceps, with Jaw Grips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72971
100488-494PK 52.55 USD InStock
100488-494
Plastic Forceps, with Jaw Grips, Electron Microscopy Sciences
Forceps
These plastic forceps are excellent for handling hot, cold, corrosive or contaminated objects. Made from blue polypropylene with interlocking teeth.
A catch locks tips tightly for use as a clamp. Scissor type handles with double action hinges makes this forcep a useful cryo tool, and great for handling stubs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty