Would you like to log in again?
Polyester Wax, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
19312
100496-094EA 371.6 USD InStock
100496-094
Polyester Wax, Electron Microscopy Sciences
Lubricants
A synthetic polyester wax with a very low melting point (37 °C). Invented by Dr. H.F. Steedman as a ribboning embedding medium. Its main advantage over paraffin and ester waxes is its low melting point, which reduces tissue hardening, shrinkage, and eliminates the need for infiltration ovens.
  • Soluble in most organic solvents, including alcohol, ethers, esters, ketones and hydrocarbons
  • Water tolerant, almost opaque, and sections easily
  • No electrification of ribbons occurs during sectioning
  • Sections of 2 microns and more may be cut at room temperatures between 10 and 22 °C
  • Advantageous to keep the wax in its solid state prior to use
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty