Would you like to log in again?
Quetol 651 - NSA, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14640
100503-462KT 287.34 USD InStock
100503-462
Quetol 651 - NSA, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Quetol 651 - NSA is a water miscible epoxy resin used for both light and electron microscopy. Its low viscosity permits easy infiltration and embedding, and yields light-colored blocks.
Delivery information: Kit consists of 225 mL Quetol 651, 450 mL NMA , 450 mL NSA, 50 mL DMP-30.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty