Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Rabbit Polyclonal Antibody (Gold, >1 nm (ultra-small))

Supplier: Electron Microscopy Sciences
25340 25341
100487-728EA 370.33 USD InStock
100487-728 100487-726
Anti-IgG Rabbit Polyclonal Antibody (Gold, >1 nm (ultra-small))
Antibodies
Prepared with subnanometer gold particles, these particles have far less influence on the adsorbed antibodies or detecting molecules, and consequently behave as though they are uncoupled. Steric hindrance is decreased compared to conventional.

Aurion Ultra Small Immunogold Reagents are prepared with subnanometer gold particles. These particles have far less influence on the adsorbed antibodies or detecting molecules, and consequently the conjugates behave as though they are uncoupled. In conjunction with the highly efficient and easy-to-use R-Gent SE-LM and SE-EM silver enhancement reagents, the Ultra Small Immunogold Reagents are the best choice for any application.Reduction of the gold particle size provides Ultra Small Immunogold Reagents with fundamentally different characteristics when compared with conjugates built around larger particles. While the Conventional Immunogold Reagents can be thought of as particles coated with proteins, Ultra Small Immunogold Reagents are proteins coated with one or more gold particles. With this structure, both the overall size of the conjugates, as well as steric hindrance are decreased.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Gold, >1 nm (ultra-small)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Sheep

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty