Would you like to log in again?
Razor Blades: Injector Type, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71991 71990
100491-884EA 105.43 USD InStock
100491-884 100491-882
Razor Blades: Injector Type, Electron Microscopy Sciences
Razors and Blades
Blades are for vibratome sectioning.
Bulk packed or in a handy dispenser. Uncoated Carbon Steel, 0.010" thick, 1 1/2" long, 5/16" wide. Stainless Steel "PTFE" Coated 0.010" thick, packed 20 blades in a dispenser.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty