Would you like to log in again?
Immunogold Reagent Kits
Supplier: Electron Microscopy Sciences
25548 25540 25550-05 25550 25540-05 25550-10 25550-06 25552 25544
100487-468KT 844.58 USD InStock
100487-468 100487-466 100503-490 100487-470 102092-352 100503-488 100503-486 100487-472 102091-024
Immunogold Reagent Kits
Protein Linking and Labeling Systems
Complete kits with the following characteristics:
  • Two non-overlapping gold particle sizes
  • Coupled to: either Goat-anti-Rabbit and Goat-anti-Mouse IgG/IgM antibodies
  • Affinity purified and cross adsorbed antibodies
  • For use with rabbit and mouse monoclonal and polyclonal primary antibodies
  • Acetylated serum albumin with increased exposure of negative charges
  • Unparalleled low background levels
  • Covering over 90% of all applications in EM
  • For single and double labeling in Electron Microscopy
EM-kit GAR-6/GAM-10/BSA-c™
1ml Goat-anti-Rabbit 6nm
1ml Goat-anti-Mouse IgG/IgM 10nm
30 ml BSA-c™ background suppressing agent
EM-kit GAM-6/GAR-10/BSA-c™
1ml Goat-anti-Mouse IgG/IgM 6nm
1ml Goat-anti-Rabbit 10nm
30 ml BSA-c™ background suppressing agent
EM-kit PrA-6/PrA-10/BSA-c™
1ml Protein A 6nm
1ml Protein A 10nm
30 ml BSA-c™ background suppressing agent
Custom EM Kit
For customization of our EM Kits you get to choose the 2 vials of your EM Grade Reagent. Supplied with the kit as well as 30 ml of BSA-c™.
Custom Ultra Small Kit
For customization of our Ultra Small Kits we now have available a make it yourself kit where as you get to choose the two vials of your Ultra Small Reagent. Also supplied with the kit is 30 ml BSA-c™ and 2x30 ml of R-Gent SE-LM.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty